The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002775.4 | NP_002766.1 | 6 - 7 | 1250 | 86 |
GenBank mRNA | AF070555.1 | - | 4 - 5 | 524 | 86 |
AF097709.1 | - | 6 - 7 | 693 | 86 | |
AK091944.1 | - | 8 - 9 | 1025 | 86 | |
AK092476.1 | - | 7 - 8 | 1031 | 86 | |
AK290089.1 | - | 6 - 7 | 741 | 86 | |
BC011352.1 | - | 7 - 8 | 1017 | 86 | |
BC031082.1 | - | 6 - 7 | 1158 | 86 | |
D87258.1 | - | 6 - 7 | 1170 | 86 | |
Y07921.1 | - | 6 - 7 | 1160 | 86 |