The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018412.3 | NP_060882.2 | 7 - 8 | 1009 | 89 |
NM_021908.2 | NP_068708.1 | 8 - 9 | 1078 | 89 | |
GenBank mRNA | AF234882.1 | - | 7 - 8 | 1008 | 89 |
AF234883.1 | - | 8 - 9 | 1077 | 89 | |
AF415174.1 | - | 5 - 6 | 436 | 89 | |
AJ277291.1 | - | 6 - 7 | 644 | 89 | |
AK093212.1 | - | 8 - 9 | 1258 | 89 | |
AK292372.1 | - | 7 - 8 | 835 | 89 | |
AK297510.1 | - | 8 - 9 | 955 | 89 | |
AK297861.1 | - | 8 - 9 | 786 | 89 | |
AK298540.1 | - | 7 - 8 | 831 | 89 | |
AY009152.1 | - | 8 - 9 | 1069 | 89 | |
AY009153.1 | - | 7 - 8 | 1000 | 89 | |
BC030954.1 | - | 8 - 9 | 814 | 89 | |
BC075855.1 | - | 8 - 9 | 1078 | 89 | |
CR936656.1 | - | 2 - 3 | 219 | 89 | |
KJ534947.1 | - | 8 - 9 | 864 | 89 |