The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170460.1 | NP_001163931.1 | 12 - 13 | 1540 | 82 |
NM_006201.4 | NP_006192.1 | 12 - 13 | 1675 | 82 | |
NM_033018.3 | NP_148978.2 | 12 - 13 | 1782 | 82 | |
XM_011543920.2 | XP_011542222.1 | 12 - 13 | 1552 | 82 | |
XM_011543921.1 | XP_011542223.1 | 13 - 14 | 1947 | 82 | |
XM_011543922.1 | XP_011542224.1 | 13 - 14 | 1935 | 82 | |
XM_011543923.1 | XP_011542225.1 | 12 - 13 | 1865 | 82 | |
XM_011543924.1 | XP_011542226.1 | 12 - 13 | 1675 | 82 | |
XM_011543925.2 | XP_011542227.1 | 13 - 14 | 1757 | 82 | |
XM_011543926.1 | XP_011542228.1 | 12 - 13 | 2718 | 82 | |
XM_011543927.2 | XP_011542229.1 | 13 - 14 | 1795 | 82 | |
XM_011543928.1 | XP_011542230.1 | 13 - 14 | 2150 | 82 | |
XM_017029569.1 | XP_016885058.1 | 13 - 14 | 1947 | 82 | |
XM_017029570.1 | XP_016885059.1 | 13 - 14 | 1757 | 82 | |
XM_017029571.1 | XP_016885060.1 | 12 - 13 | 2718 | 82 | |
XM_017029572.1 | XP_016885061.1 | 13 - 14 | 1795 | 82 | |
XM_017029573.1 | XP_016885062.1 | 13 - 14 | 2150 | 82 | |
GenBank mRNA | AB209465.1 | - | 9 - 10 | 1912 | 82 |
AK096881.1 | - | 12 - 13 | 1664 | 82 | |
AK290145.1 | - | 12 - 13 | 1599 | 82 | |
AK301832.1 | - | 12 - 13 | 1540 | 82 | |
BC001048.2 | - | 12 - 13 | 1678 | 82 | |
BC006190.2 | - | 12 - 13 | 1492 | 82 | |
BC009852.2 | - | 10 - 11 | 938 | 82 | |
BC015607.2 | - | 12 - 13 | 1645 | 82 | |
BT006827.1 | - | 11 - 12 | 1240 | 82 | |
X66363.1 | - | 12 - 13 | 1363 | 82 |