The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204466.1 | NP_001191395.1 | 12 - 13 | 1598 | 68 |
NM_001204467.1 | NP_001191396.1 | 13 - 14 | 1826 | 68 | |
NM_001204468.1 | NP_001191397.1 | 13 - 14 | 1860 | 68 | |
NM_005676.4 | NP_005667.2 | 13 - 14 | 1829 | 68 | |
NM_152856.2 | NP_690595.1 | 12 - 13 | 1595 | 68 | |
XM_005272677.4 | XP_005272734.1 | 13 - 14 | 1879 | 68 | |
XM_005272678.4 | XP_005272735.1 | 12 - 13 | 1413 | 68 | |
XM_005272679.4 | XP_005272736.1 | 12 - 13 | 1410 | 68 | |
XM_006724563.1 | XP_006724626.1 | 9 - 10 | 1046 | 68 | |
XM_011543989.2 | XP_011542291.1 | 5 - 6 | 770 | 68 | |
XM_017029884.1 | XP_016885373.1 | 12 - 13 | 1749 | 68 | |
XM_017029885.1 | XP_016885374.1 | 12 - 13 | 1521 | 68 | |
GenBank mRNA | AK098115.1 | - | - | 2438 | 68 |
AK292758.1 | - | 12 - 13 | 1549 | 68 | |
AL137421.1 | - | 14 - 15 | 1718 | 68 | |
BC000681.2 | - | 10 - 11 | 1051 | 68 | |
BC003089.1 | - | 12 - 13 | 1534 | 68 | |
BC004181.2 | - | 13 - 14 | 1782 | 68 | |
BC008733.2 | - | 13 - 14 | 1814 | 68 | |
BC024153.2 | - | 12 - 13 | 1444 | 68 | |
BX537969.1 | - | 13 - 14 | 1811 | 68 | |
D50912.1 | - | 13 - 14 | 1677 | 68 | |
KU178411.1 | - | 11 - 12 | 1202 | 68 | |
KU178412.1 | - | 12 - 13 | 1436 | 68 |