The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032775.3 | NP_116164.2 | 5 - 6 | 1464 | 93 |
NR_033825.1 | 4 - 5 | 1298 | 93 | ||
XM_017029018.1 | XP_016884507.1 | 4 - 5 | 1632 | 93 | |
XM_017029019.1 | XP_016884508.1 | 5 - 6 | 1760 | 93 | |
XM_017029020.1 | XP_016884509.1 | 6 - 7 | 2939 | 93 | |
XM_017029021.1 | XP_016884510.1 | 5 - 6 | 1393 | 93 | |
XM_017029022.1 | XP_016884511.1 | 4 - 5 | 1564 | 93 | |
XM_017029023.1 | XP_016884512.1 | 4 - 5 | 4024 | 93 | |
XM_017029024.1 | XP_016884513.1 | 4 - 5 | 1596 | 93 | |
XM_017029025.1 | XP_016884514.1 | 4 - 5 | 1112 | 93 | |
GenBank mRNA | AK027266.1 | - | 5 - 6 | 1403 | 93 |
AK222850.1 | - | 5 - 6 | 1370 | 93 | |
AK290669.1 | - | 5 - 6 | 1376 | 93 | |
AK294682.1 | - | 4 - 5 | 1210 | 93 | |
BC015923.1 | - | 5 - 6 | 1416 | 93 | |
CR456352.1 | - | 4 - 5 | 1341 | 93 |