The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014718.3 | NP_055533.2 | 15 - 16 | 2672 | 67 |
XM_006719163.3 | XP_006719226.1 | 14 - 15 | 3998 | 67 | |
GenBank mRNA | AB018269.1 | - | 14 - 15 | 2968 | 67 |
AJ277460.1 | - | 15 - 16 | 2669 | 67 | |
AK299177.1 | - | 15 - 16 | 2646 | 67 | |
AK299246.1 | - | 6 - 7 | 874 | 67 | |
AK316059.1 | - | 15 - 16 | 2672 | 67 | |
AY753302.1 | - | 14 - 15 | 2430 | 67 | |
BC104767.1 | - | 15 - 16 | 2476 | 67 | |
BC111491.1 | - | 15 - 16 | 2660 | 67 | |
BC112283.1 | - | 15 - 16 | 2488 | 67 |