Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCAN
Entrez Gene ID
63827
Gene Name
brevican
Gene Aliases
BEHAB, CSPG7
Chromosome Location
Chr.1: 156641948 - 156659532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021948.4 NP_068767.3 9 - 10 2390 111
XM_011509866.1 XP_011508168.1 9 - 10 2217 111
XM_017002047.1 XP_016857536.1 9 - 10 2311 111
GenBank mRNA AB209570.1 - 7 - 8 1872 111
AF228710.1 - 9 - 10 2101 111
AK289759.1 - 9 - 10 2221 111
AL137504.1 - 2 - 3 311 111
AY358372.1 - 9 - 10 2405 111
BC009117.1 - 9 - 10 2178 111
BC010571.1 - 9 - 10 2156 111
BC013234.1 - 2 - 3 359 111
BC022938.1 - 9 - 10 2202 111
BC023209.1 - 5 - 6 1369 111
BC027971.1 - 9 - 10 2194 111
BC029313.1 - 7 - 8 1674 111
BC035457.1 - 9 - 10 2185 111

View Full Product Details