The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195248.1 | NP_001182177.1 | 3 - 4 | 259 | 78 |
NM_001195249.1 | NP_001182178.1 | 3 - 4 | 428 | 78 | |
NM_001195251.1 | NP_001182180.1 | 4 - 5 | 365 | 78 | |
NM_001195252.1 | NP_001182181.1 | 3 - 4 | 259 | 78 | |
NM_175069.2 | NP_778239.1 | 3 - 4 | 259 | 78 | |
NM_175073.2 | NP_778243.1 | 4 - 5 | 365 | 78 | |
NR_036576.1 | 3 - 4 | 254 | 78 | ||
NR_036578.1 | 3 - 4 | 276 | 78 | ||
NR_036579.1 | 3 - 4 | 423 | 78 | ||
XM_006716791.3 | XP_006716854.1 | 3 - 4 | 267 | 78 | |
XM_006716792.2 | XP_006716855.1 | 3 - 4 | 285 | 78 | |
XM_017014831.1 | XP_016870320.1 | 3 - 4 | 259 | 78 | |
XM_017014832.1 | XP_016870321.1 | 3 - 4 | 259 | 78 | |
XM_017014833.1 | XP_016870322.1 | 3 - 4 | 259 | 78 | |
XM_017014834.1 | XP_016870323.1 | 3 - 4 | 259 | 78 | |
XM_017014835.1 | XP_016870324.1 | 3 - 4 | 259 | 78 | |
XM_017014836.1 | XP_016870325.1 | 3 - 4 | 259 | 78 | |
XM_017014838.1 | XP_016870327.1 | 3 - 4 | 241 | 78 | |
GenBank mRNA | AB603741.1 | - | 2 - 3 | 155 | 78 |
AJ565852.1 | - | 3 - 4 | 232 | 78 | |
AJ565853.1 | - | 2 - 3 | 235 | 78 | |
AJ565854.1 | - | 2 - 3 | 240 | 78 | |
AK055672.1 | - | 3 - 4 | 246 | 78 | |
AK131046.1 | - | 3 - 4 | 211 | 78 | |
AY040777.1 | - | 3 - 4 | 234 | 78 | |
AY208829.1 | - | 3 - 4 | 234 | 78 | |
AY208831.1 | - | 3 - 4 | 225 | 78 | |
AY208834.1 | - | 3 - 4 | 230 | 78 | |
AY208835.1 | - | 3 - 4 | 214 | 78 | |
AY208836.1 | - | 3 - 4 | 374 | 78 | |
AY208837.1 | - | 4 - 5 | 331 | 78 | |
AY208839.1 | - | 3 - 4 | 368 | 78 | |
AY208840.1 | - | 3 - 4 | 231 | 78 | |
AY302067.1 | - | 2 - 3 | 377 | 78 | |
AY302068.1 | - | 2 - 3 | 372 | 78 | |
AY302071.1 | - | 2 - 3 | 377 | 78 | |
AY302072.1 | - | 2 - 3 | 372 | 78 | |
BC032650.1 | - | 3 - 4 | 300 | 78 | |
BC068107.1 | - | 4 - 5 | 340 | 78 | |
BC104881.1 | - | 3 - 4 | 273 | 78 | |
BQ957536.1 | - | 4 - 5 | 348 | 78 | |
BX538161.1 | - | 4 - 5 | 344 | 78 | |
DA664623.1 | - | 2 - 3 | 286 | 78 |