The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040444.2 | NP_001035534.1 | 1 - 2 | 158 | 108 |
NR_135323.1 | 2 - 3 | 294 | 108 | ||
GenBank mRNA | AF155105.1 | - | 1 - 2 | 101 | 108 |
AK225986.1 | - | 2 - 3 | 244 | 108 | |
DB050568.1 | - | 2 - 3 | 249 | 108 |