The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319116.1 | NP_001306045.1 | 11 - 12 | 1223 | 84 |
NM_001319117.1 | NP_001306046.1 | 10 - 11 | 1115 | 84 | |
NM_001319118.1 | NP_001306047.1 | 11 - 12 | 1151 | 84 | |
NM_001319119.1 | NP_001306048.1 | 11 - 12 | 1431 | 84 | |
NM_001319120.1 | NP_001306049.1 | 10 - 11 | 1140 | 84 | |
NM_001319121.1 | NP_001306050.1 | 12 - 13 | 1124 | 84 | |
NM_001319122.1 | NP_001306051.1 | 11 - 12 | 1216 | 84 | |
NM_012100.3 | NP_036232.2 | 11 - 12 | 1248 | 84 | |
NR_134970.1 | 9 - 10 | 1064 | 84 | ||
GenBank mRNA | AF005050.2 | - | 11 - 12 | 1223 | 84 |
AK001777.1 | - | 11 - 12 | 1248 | 84 | |
AK001977.1 | - | 11 - 12 | 1220 | 84 | |
AK023005.1 | - | 11 - 12 | 1162 | 84 | |
BC000653.2 | - | 11 - 12 | 1130 | 84 | |
BC004854.2 | - | 7 - 8 | 781 | 84 |