The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290292.1 | NP_001277221.1 | 2 - 3 | 199 | 102 |
NM_001290293.2 | NP_001277222.1 | 2 - 3 | 161 | 102 | |
NM_001308075.1 | NP_001295004.1 | 2 - 3 | 161 | 102 | |
NM_020195.2 | NP_064580.2 | 2 - 3 | 159 | 102 | |
XM_011536975.1 | XP_011535277.1 | 2 - 3 | 134 | 102 | |
GenBank mRNA | AF226050.1 | - | 1 - 2 | 433 | 102 |
BC000989.2 | - | 2 - 3 | 159 | 102 | |
BC008566.1 | - | 1 - 2 | 331 | 102 | |
BX161384.1 | - | 1 - 2 | 112 | 102 | |
CN406892.1 | - | 1 - 2 | 123 | 102 | |
CR995681.1 | - | 1 - 2 | 125 | 102 | |
DT220226.1 | - | 2 - 3 | 102 |