Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPTN
Entrez Gene ID
10133
Gene Name
optineurin
Gene Aliases
ALS12, FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP
Chromosome Location
Chr.10: 13100082 - 13138276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021980.4 NP_068815.2 1 - 2 225 85
GenBank mRNA AF420372.1 - 1 - 2 82 85
AK303715.1 - 1 - 2 149 85
AK316171.1 - 1 - 2 171 85
BC013876.2 - 1 - 2 138 85

View Full Product Details