Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC13
Entrez Gene ID
79573
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
-
Chromosome Location
Chr.1: 230906241 - 230978872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122835.2 NP_001116307.2 18 - 19 2184 106
NM_024525.4 NP_078801.3 20 - 21 2346 106
XM_005273260.1 XP_005273317.1 20 - 21 2343 106
XM_005273261.2 XP_005273318.1 19 - 20 2253 106
XM_005273262.1 XP_005273319.1 18 - 19 2187 106
XM_005273264.2 XP_005273321.1 18 - 19 2118 106
XM_006711814.2 XP_006711877.1 16 - 17 1959 106
XM_006711815.2 XP_006711878.1 13 - 14 3353 106
XM_011544275.2 XP_011542577.1 17 - 18 2025 106
XM_011544276.2 XP_011542578.1 13 - 14 1589 106
XM_017002321.1 XP_016857810.1 19 - 20 2250 106
XM_017002322.1 XP_016857811.1 18 - 19 2115 106
XM_017002323.1 XP_016857812.1 16 - 17 1921 106
XM_017002324.1 XP_016857813.1 16 - 17 1956 106
XM_017002325.1 XP_016857814.1 20 - 21 5548 106
XM_017002326.1 XP_016857815.1 15 - 16 4928 106
GenBank mRNA AK026237.1 - 7 - 8 653 106
AK074365.1 - 11 - 12 1254 106
AK075386.1 - 20 - 21 2312 106
AK075561.1 - 18 - 19 2163 106
AK309725.1 - 16 - 17 2002 106
AL832286.1 - 20 - 21 2081 106
AL834453.1 - 11 - 12 1312 106
BC042683.1 - 11 - 12 1230 106

View Full Product Details