The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122835.2 | NP_001116307.2 | 2 - 3 | 410 | 71 |
NM_024525.4 | NP_078801.3 | 2 - 3 | 410 | 71 | |
XM_005273260.1 | XP_005273317.1 | 2 - 3 | 410 | 71 | |
XM_005273261.2 | XP_005273318.1 | 2 - 3 | 410 | 71 | |
XM_005273262.1 | XP_005273319.1 | 2 - 3 | 410 | 71 | |
XM_005273264.2 | XP_005273321.1 | 2 - 3 | 410 | 71 | |
XM_006711814.2 | XP_006711877.1 | 2 - 3 | 410 | 71 | |
XM_011544275.2 | XP_011542577.1 | 2 - 3 | 410 | 71 | |
XM_017002321.1 | XP_016857810.1 | 2 - 3 | 410 | 71 | |
XM_017002322.1 | XP_016857811.1 | 2 - 3 | 410 | 71 | |
XM_017002323.1 | XP_016857812.1 | 2 - 3 | 372 | 71 | |
XM_017002324.1 | XP_016857813.1 | 2 - 3 | 410 | 71 | |
XM_017002325.1 | XP_016857814.1 | 2 - 3 | 387 | 71 | |
GenBank mRNA | AK075386.1 | - | 2 - 3 | 376 | 71 |
AK075561.1 | - | 2 - 3 | 389 | 71 | |
AL832286.1 | - | 2 - 3 | 145 | 71 | |
DA468037.1 | - | 2 - 3 | 392 | 71 | |
DA741498.1 | - | 2 - 3 | 410 | 71 |