The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004860.3 | NP_004851.2 | 3 - 4 | 588 | 86 |
GenBank mRNA | AK313836.1 | - | 3 - 4 | 273 | 86 |
BC020090.1 | - | 3 - 4 | 542 | 86 | |
BC051907.1 | - | 3 - 4 | 419 | 86 | |
BC067272.1 | - | 3 - 4 | 533 | 86 | |
BT009817.1 | - | 3 - 4 | 223 | 86 | |
U31501.1 | - | 3 - 4 | 450 | 86 |