The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001715.2 | NP_001706.2 | 3 - 4 | 759 | 76 |
XM_011543824.1 | XP_011542126.1 | 3 - 4 | 780 | 76 | |
XM_011543825.2 | XP_011542127.1 | 3 - 4 | 525 | 76 | |
XM_011543827.1 | XP_011542129.1 | 2 - 3 | 656 | 76 | |
XM_011543828.2 | XP_011542130.1 | 3 - 4 | 780 | 76 | |
XM_011543829.2 | XP_011542131.1 | 3 - 4 | 780 | 76 | |
XM_017013730.1 | XP_016869219.1 | 2 - 3 | 656 | 76 | |
GenBank mRNA | AK313751.1 | - | 3 - 4 | 348 | 76 |
BC007371.2 | - | 3 - 4 | 322 | 76 | |
BC018703.1 | - | 3 - 4 | 623 | 76 | |
BC032413.1 | - | 3 - 4 | 368 | 76 | |
S76617.1 | - | 3 - 4 | 766 | 76 | |
Z33998.1 | - | 3 - 4 | 400 | 76 |