Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL5A
Entrez Gene ID
26225
Gene Name
ADP ribosylation factor like GTPase 5A
Gene Aliases
ARFLP5, ARL5
Chromosome Location
Chr.2: 151800966 - 151828495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037174.1 NP_001032251.1 2 - 3 172 138
NM_012097.3 NP_036229.1 2 - 3 428 138
NM_177985.2 NP_817114.2 2 - 3 170 138
GenBank mRNA AF087889.1 - - 358 138
AF100740.1 - 2 - 3 175 138
AF493891.1 - 2 - 3 109 138
AK292714.1 - 2 - 3 338 138
AK298937.1 - 2 - 3 116 138
BC001254.1 - 2 - 3 346 138
BG707385.1 - 2 - 3 149 138
BQ216648.1 - 2 - 3 415 138
BT009829.1 - 2 - 3 109 138
BX461356.2 - 2 - 3 143 138
CX867116.1 - 2 - 3 434 138

View Full Product Details