Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
Chromosome Location
Chr.7: 14145049 - 14974777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004080.2 NP_004071.1 19 - 20 1961 79
NM_145695.2 NP_663733.1 19 - 20 1961 79
XM_005249628.2 XP_005249685.1 20 - 21 2489 79
XM_005249629.2 XP_005249686.1 20 - 21 2486 79
XM_005249630.2 XP_005249687.1 19 - 20 2002 79
XM_005249631.3 XP_005249688.1 18 - 19 2217 79
XM_011515153.2 XP_011513455.1 20 - 21 2327 79
XM_011515154.2 XP_011513456.1 20 - 21 2043 79
XM_011515156.2 XP_011513458.1 19 - 20 1999 79
XM_011515157.2 XP_011513459.1 19 - 20 2006 79
XM_011515158.2 XP_011513460.1 18 - 19 2142 79
XM_017011783.1 XP_016867272.1 20 - 21 2380 79
XM_017011784.1 XP_016867273.1 20 - 21 2040 79
XM_017011785.1 XP_016867274.1 19 - 20 2274 79
XM_017011786.1 XP_016867275.1 19 - 20 2254 79
XM_017011787.1 XP_016867276.1 19 - 20 2004 79
XM_017011788.1 XP_016867277.1 19 - 20 2290 79
XM_017011789.1 XP_016867278.1 19 - 20 2007 79
XM_017011790.1 XP_016867279.1 20 - 21 2326 79
XM_017011791.1 XP_016867280.1 20 - 21 2325 79
XM_017011792.1 XP_016867281.1 20 - 21 2325 79
GenBank mRNA AB018261.1 - 16 - 17 1590 79
AK304719.1 - 18 - 19 2217 79
BC105005.1 - 19 - 20 1841 79
BC143632.1 - 19 - 20 1838 79
BC143633.1 - 18 - 19 1800 79

View Full Product Details