The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372.3 | NP_001363.2 | 60 - 61 | 11672 | 65 |
NM_004662.2 | NP_004653.2 | 6 - 7 | 1069 | 65 | |
XM_011523703.1 | XP_011522005.1 | 60 - 61 | 11636 | 65 | |
XM_017024292.1 | XP_016879781.1 | 59 - 60 | 11519 | 65 | |
XM_017024293.1 | XP_016879782.1 | 49 - 50 | 9580 | 65 | |
GenBank mRNA | AB002355.2 | - | 41 - 42 | 7350 | 65 |
AB290158.1 | - | 48 - 49 | 9489 | 65 | |
AF015265.1 | - | 1 - 2 | 117 | 65 | |
AF257737.1 | - | 60 - 61 | 11662 | 65 | |
AJ404468.1 | - | 60 - 61 | 11672 | 65 | |
BC128261.1 | - | 4 - 5 | 609 | 65 | |
BC128421.1 | - | 4 - 5 | 609 | 65 | |
BC153883.1 | - | 41 - 42 | 7350 | 65 | |
X99947.1 | - | 6 - 7 | 1069 | 65 |