Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
Chromosome Location
Chr.17: 11598431 - 11970168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372.3 NP_001363.2 3 - 4 845 84
XM_011523703.1 XP_011522005.1 3 - 4 809 84
XM_017024292.1 XP_016879781.1 3 - 4 809 84
XM_017024294.1 XP_016879783.1 3 - 4 809 84
XM_017024295.1 XP_016879784.1 3 - 4 809 84
GenBank mRNA AF257737.1 - 3 - 4 835 84
AJ404468.1 - 3 - 4 845 84
AK055990.1 - 3 - 4 807 84

View Full Product Details