The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015261.2 | NP_056076.1 | 32 - 33 | 4862 | 59 |
XM_011542719.2 | XP_011541021.1 | 31 - 32 | 4123 | 59 | |
GenBank mRNA | AF070553.1 | - | 6 - 7 | 775 | 59 |
AK025549.1 | - | 6 - 7 | 892 | 59 | |
AK025667.1 | - | 16 - 17 | 2153 | 59 | |
AK124878.1 | - | 32 - 33 | 4788 | 59 | |
BC011408.1 | - | 10 - 11 | 1437 | 59 | |
BC098398.1 | - | 32 - 33 | 4321 | 59 | |
BC144524.1 | - | 33 - 34 | 4687 | 59 | |
D29954.1 | - | 32 - 33 | 4284 | 59 |