The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015261.2 | NP_056076.1 | 2 - 3 | 820 | 92 |
GenBank mRNA | AK124878.1 | - | 2 - 3 | 820 | 92 |
BC098398.1 | - | 2 - 3 | 279 | 92 | |
BC144524.1 | - | 2 - 3 | 504 | 92 | |
D29954.1 | - | 2 - 3 | 242 | 92 |