The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032974.4 | NP_116756.2 | 6 - 7 | 1141 | 62 |
NM_032977.3 | NP_116759.2 | 6 - 7 | 1141 | 62 | |
XM_005246907.2 | XP_005246964.1 | 6 - 7 | 924 | 62 | |
GenBank mRNA | AF111345.1 | - | 5 - 6 | 723 | 62 |
AK310746.1 | - | 6 - 7 | 927 | 62 | |
BC042844.1 | - | 6 - 7 | 877 | 62 | |
KU177959.1 | - | 5 - 6 | 723 | 62 | |
U86214.1 | - | 6 - 7 | 917 | 62 |