The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002926.3 | NP_002917.1 | 2 - 3 | 2787 | 113 |
NM_198229.2 | NP_937872.1 | 2 - 3 | 2787 | 113 | |
XM_006713905.1 | XP_006713968.1 | 2 - 3 | 2106 | 113 | |
XM_006713906.2 | XP_006713969.1 | 2 - 3 | 2106 | 113 | |
XM_017008529.1 | XP_016864018.1 | 2 - 3 | 3356 | 113 | |
XM_017008530.1 | XP_016864019.1 | 2 - 3 | 2106 | 113 | |
XM_017008531.1 | XP_016864020.1 | 1 - 2 | 1303 | 113 | |
GenBank mRNA | AF030111.1 | - | 2 - 3 | 2786 | 113 |
AF035152.1 | - | 1 - 2 | 1937 | 113 | |
AF464735.1 | - | 1 - 2 | 1986 | 113 | |
AF464736.1 | - | 1 - 2 | 1986 | 113 | |
AK293767.1 | - | 2 - 3 | 2039 | 113 | |
BC028154.1 | - | 2 - 3 | 2049 | 113 |