The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024844.1 | NP_001020015.1 | 6 - 7 | 610 | 74 |
NM_002231.3 | NP_002222.1 | 7 - 8 | 685 | 74 | |
XM_006718223.2 | XP_006718286.1 | 8 - 9 | 724 | 74 | |
XM_011520067.2 | XP_011518369.1 | 4 - 5 | 387 | 74 | |
GenBank mRNA | AK225410.1 | - | 7 - 8 | 611 | 74 |
AK225708.1 | - | 7 - 8 | 639 | 74 | |
AK312677.1 | - | 7 - 8 | 611 | 74 | |
AY303776.1 | - | 4 - 5 | 362 | 74 | |
BC000726.2 | - | 7 - 8 | 583 | 74 | |
BC001821.2 | - | 7 - 8 | 583 | 74 | |
BT019761.1 | - | 5 - 6 | 437 | 74 | |
BT019762.1 | - | 5 - 6 | 437 | 74 | |
CR407650.1 | - | 5 - 6 | 437 | 74 | |
CR542255.1 | - | 5 - 6 | 437 | 74 | |
S48196.1 | - | 7 - 8 | 603 | 74 | |
U20770.1 | - | 7 - 8 | 602 | 74 |