Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTPD6
Entrez Gene ID
955
Gene Name
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
Chromosome Location
Chr.20: 25195670 - 25228075 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114089.3 NP_001107561.1 8 - 9 1024 70
NM_001247.4 NP_001238.2 9 - 10 1093 70
NM_001317941.2 NP_001304870.1 9 - 10 1090 70
NM_001322378.1 NP_001309307.1 9 - 10 1237 70
NM_001322379.1 NP_001309308.1 7 - 8 702 70
NM_001322380.1 NP_001309309.1 7 - 8 702 70
NM_001322381.1 NP_001309310.1 6 - 7 558 70
NM_001322382.1 NP_001309311.1 6 - 7 558 70
NM_001322383.1 NP_001309312.1 10 - 11 1281 70
NM_001322384.1 NP_001309313.1 7 - 8 702 70
NM_001322385.1 NP_001309314.1 7 - 8 880 70
NM_001322386.1 NP_001309315.1 8 - 9 1024 70
NM_001322387.1 NP_001309316.1 8 - 9 949 70
NM_001322388.1 NP_001309317.1 9 - 10 1093 70
NM_001322389.1 NP_001309318.1 8 - 9 1168 70
NM_001322390.1 NP_001309319.1 10 - 11 1215 70
NM_001322391.1 NP_001309320.1 9 - 10 1093 70
NM_001322392.1 NP_001309321.1 8 - 9 1021 70
NM_001322393.1 NP_001309322.1 8 - 9 1024 70
NM_001322394.1 NP_001309323.1 8 - 9 1021 70
NM_001322395.1 NP_001309324.1 8 - 9 1168 70
NM_001322396.1 NP_001309325.1 9 - 10 1090 70
NM_001322397.1 NP_001309326.1 9 - 10 1093 70
NM_001322398.1 NP_001309327.1 8 - 9 1168 70
XM_006723665.2 XP_006723728.1 10 - 11 1129 70
XM_011529396.1 XP_011527698.1 10 - 11 1133 70
XM_011529397.1 XP_011527699.1 10 - 11 1062 70
XM_017028129.1 XP_016883618.1 8 - 9 902 70
XM_017028130.1 XP_016883619.1 7 - 8 758 70
XM_017028131.1 XP_016883620.1 8 - 9 902 70
XM_017028132.1 XP_016883621.1 8 - 9 925 70
XM_017028133.1 XP_016883622.1 8 - 9 872 70
XM_017028134.1 XP_016883623.1 7 - 8 669 70
GenBank mRNA AF039916.1 - 9 - 10 1027 70
AK074691.1 - 8 - 9 873 70
AK293256.1 - 8 - 9 965 70
AK295242.1 - 4 - 5 274 70
AK297954.1 - 9 - 10 933 70
AK300028.1 - 7 - 8 653 70
AK302395.1 - 9 - 10 941 70
AK303786.1 - 8 - 9 979 70
AK308626.1 - 8 - 9 991 70
AL834158.1 - 8 - 9 939 70
AY327581.1 - 9 - 10 990 70
BC025980.2 - 9 - 10 70
CR936765.1 - 9 - 10 1007 70

View Full Product Details