The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014396.3 | NP_055211.2 | 20 - 21 | 1762 | 78 |
NM_080631.3 | NP_542198.2 | 19 - 20 | 1687 | 78 | |
XM_017011988.1 | XP_016867477.1 | 7 - 8 | 695 | 78 | |
GenBank mRNA | AK074325.1 | - | 16 - 17 | 1475 | 78 |
AK299143.1 | - | 17 - 18 | 1508 | 78 | |
AK314556.1 | - | 20 - 21 | 1762 | 78 | |
BC044851.1 | - | 20 - 21 | 1757 | 78 | |
BX648347.1 | - | 18 - 19 | 1487 | 78 | |
U87309.1 | - | 20 - 21 | 1765 | 78 |