The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199162.2 | NP_954631.1 | 1 - 2 | 142 | 78 |
GenBank mRNA | BC009755.2 | - | 1 - 2 | 142 | 78 |