Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIK3IP1
Entrez Gene ID
113791
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
HGFL, hHGFL(S)
Chromosome Location
Chr.22: 31281593 - 31292534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135911.1 NP_001129383.1 2 - 3 380 89
NM_052880.4 NP_443112.2 2 - 3 380 89
GenBank mRNA AF528079.1 - 2 - 3 325 89
AF528080.1 - 2 - 3 325 89
AK074688.1 - 2 - 3 326 89
AK226089.1 - 2 - 3 260 89
AK299397.1 - 2 - 3 334 89
AK300990.1 - 2 - 3 304 89
BC011049.2 - 2 - 3 325 89
BQ718653.1 - 2 - 3 358 89
CR456340.1 - 2 - 3 220 89
DC412134.1 - 2 - 3 380 89

View Full Product Details