Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIK3IP1
Entrez Gene ID
113791
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
HGFL, hHGFL(S)
Chromosome Location
Chr.22: 31281593 - 31292534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052880.4 NP_443112.2 4 - 5 701 89
GenBank mRNA AF528079.1 - 4 - 5 646 89
AF528080.1 - 4 - 5 646 89
AK093768.1 - 1 - 2 1261 89
AK226089.1 - 4 - 5 581 89
AK300990.1 - 5 - 6 559 89
BC011049.2 - 4 - 5 646 89
BC041903.1 - 2 - 3 998 89
BE299406.1 - 2 - 3 329 89
BQ718653.1 - 4 - 5 679 89
CR456340.1 - 4 - 5 541 89
GU135609.1 - 2 - 3 325 89

View Full Product Details