The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152743.3 | NP_689956.2 | 13 - 14 | 2058 | 81 |
XM_011515181.2 | XP_011513483.1 | 14 - 15 | 2100 | 81 | |
XM_017011834.1 | XP_016867323.1 | 13 - 14 | 2012 | 81 | |
XM_017011835.1 | XP_016867324.1 | 12 - 13 | 1905 | 81 | |
GenBank mRNA | AK131083.1 | - | 10 - 11 | 4361 | 81 |
AL133088.1 | - | 6 - 7 | 735 | 81 | |
BC007209.1 | - | 9 - 10 | 1293 | 81 | |
BC015632.2 | - | 13 - 14 | 1812 | 81 | |
BC023561.2 | - | 9 - 10 | 1053 | 81 |