Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPRIN3
Entrez Gene ID
285513
Gene Name
GPRIN family member 3
Gene Aliases
GRIN3
Chromosome Location
Chr.4: 89237666 - 89308010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB095947.2 - 1 - 1 961 104

View Full Product Details