Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP28
Entrez Gene ID
57646
Gene Name
ubiquitin specific peptidase 28
Gene Aliases
-
Chromosome Location
Chr.11: 113797875 - 113875580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301029.1 NP_001287958.1 5 - 6 617 84
NM_020886.3 NP_065937.1 7 - 8 828 84
XM_005271630.2 XP_005271687.1 7 - 8 824 84
XM_005271631.2 XP_005271688.1 7 - 8 824 84
XM_005271632.2 XP_005271689.1 8 - 9 1154 84
XM_005271633.4 XP_005271690.1 8 - 9 882 84
XM_005271636.2 XP_005271693.1 7 - 8 824 84
XM_005271637.2 XP_005271694.1 7 - 8 824 84
XM_005271638.2 XP_005271695.1 7 - 8 824 84
XM_005271639.2 XP_005271696.1 7 - 8 824 84
XM_011542936.2 XP_011541238.1 6 - 7 743 84
XM_011542938.1 XP_011541240.1 5 - 6 617 84
XM_017018056.1 XP_016873545.1 6 - 7 743 84
XM_017018057.1 XP_016873546.1 8 - 9 1154 84
XM_017018058.1 XP_016873547.1 6 - 7 760 84
XM_017018059.1 XP_016873548.1 6 - 7 743 84
XM_017018060.1 XP_016873549.1 5 - 6 617 84
XM_017018061.1 XP_016873550.1 5 - 6 617 84
XM_017018062.1 XP_016873551.1 5 - 6 617 84
XM_017018065.1 XP_016873554.1 7 - 8 792 84
XM_017018066.1 XP_016873555.1 8 - 9 1154 84
GenBank mRNA AF266283.1 - 7 - 8 765 84
AK304770.1 - 5 - 6 588 84
BC065928.1 - 7 - 8 786 84

View Full Product Details