The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126131.1 | NP_001119603.1 | 22 - 23 | 3917 | 119 |
NM_002693.2 | NP_002684.1 | 22 - 23 | 3929 | 119 | |
GenBank mRNA | AK130624.1 | - | 5 - 6 | 803 | 119 |
AK291281.1 | - | 22 - 23 | 3948 | 119 | |
BC042571.1 | - | 22 - 23 | 3914 | 119 | |
BC050559.1 | - | 22 - 23 | 3898 | 119 | |
D84103.1 | - | 21 - 22 | 3753 | 119 | |
U60325.1 | - | 21 - 22 | 3806 | 119 | |
X98093.1 | - | 22 - 23 | 3929 | 119 |