Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBL3
Entrez Gene ID
5412
Gene Name
ubiquitin like 3
Gene Aliases
HCG-1, PNSC1
Chromosome Location
Chr.13: 29764408 - 29850683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007106.3 NP_009037.1 1 - 2 1166 110
GenBank mRNA AF044221.1 - 1 - 2 125 110
AK056580.1 - 1 - 2 1061 110
AK311846.1 - 1 - 2 209 110
AL080177.1 - 1 - 2 130 110
BC059385.1 - 1 - 2 506 110

View Full Product Details