The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031898.2 | NP_114104.1 | 6 - 7 | 1060 | 96 |
XM_011523988.2 | XP_011522290.1 | 6 - 7 | 955 | 96 | |
XM_011523989.1 | XP_011522291.1 | 5 - 6 | 1017 | 96 | |
XM_011523990.2 | XP_011522292.1 | 7 - 8 | 2377 | 96 | |
XM_017024954.1 | XP_016880443.1 | 6 - 7 | 1104 | 96 | |
XM_017024955.1 | XP_016880444.1 | 6 - 7 | 800 | 96 | |
GenBank mRNA | AF334676.1 | - | 5 - 6 | 972 | 96 |
AK057390.1 | - | 6 - 7 | 1060 | 96 | |
AK223534.1 | - | 6 - 7 | 1016 | 96 | |
AK314327.1 | - | 6 - 7 | 1052 | 96 | |
BC031688.1 | - | 6 - 7 | 1023 | 96 |