The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031898.2 | NP_114104.1 | 8 - 9 | 1442 | 78 |
XM_011523988.2 | XP_011522290.1 | 8 - 9 | 1337 | 78 | |
XM_011523989.1 | XP_011522291.1 | 7 - 8 | 1399 | 78 | |
XM_011523990.2 | XP_011522292.1 | 9 - 10 | 2759 | 78 | |
XM_017024954.1 | XP_016880443.1 | 8 - 9 | 1486 | 78 | |
XM_017024955.1 | XP_016880444.1 | 8 - 9 | 1182 | 78 | |
GenBank mRNA | AF334676.1 | - | 7 - 8 | 1354 | 78 |
AK057390.1 | - | 8 - 9 | 1442 | 78 | |
AK223534.1 | - | 8 - 9 | 1398 | 78 | |
AK314327.1 | - | 8 - 9 | 1434 | 78 | |
BC031688.1 | - | 8 - 9 | 1405 | 78 |