The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164617.1 | NP_001158089.1 | 6 - 7 | 1558 | 61 |
NM_001164618.1 | NP_001158090.1 | 5 - 6 | 1441 | 61 | |
NM_001164619.1 | NP_001158091.1 | 4 - 5 | 1327 | 61 | |
NM_004484.3 | NP_004475.1 | 5 - 6 | 1489 | 61 | |
GenBank mRNA | AK300168.1 | - | 5 - 6 | 1412 | 61 |
AK310196.1 | - | 4 - 5 | 1156 | 61 | |
BC035972.1 | - | 5 - 6 | 1437 | 61 | |
JQ943686.1 | - | 5 - 6 | 1292 | 61 | |
L47125.1 | - | 5 - 6 | 1443 | 61 | |
L47176.1 | - | 5 - 6 | 1438 | 61 | |
U50410.1 | - | 5 - 6 | 1436 | 61 | |
Z37987.1 | - | 5 - 6 | 1400 | 61 |