The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006480.4 | NP_006471.2 | 12 - 13 | 1519 | 68 |
XM_005265794.4 | XP_005265851.1 | 12 - 13 | 1689 | 68 | |
XM_005265795.4 | XP_005265852.1 | 12 - 13 | 1694 | 68 | |
GenBank mRNA | AF037195.1 | - | 8 - 9 | 951 | 68 |
AK098150.1 | - | 5 - 6 | 1444 | 68 | |
AK123382.1 | - | 8 - 9 | 863 | 68 | |
AY987041.1 | - | 8 - 9 | 881 | 68 | |
BC014094.2 | - | 12 - 13 | 1489 | 68 |