Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC13
Entrez Gene ID
79573
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
-
Chromosome Location
Chr.1: 230906241 - 230978872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006711815.2 XP_006711878.1 1 - 2 1951 147
XM_017002325.1 XP_016857814.1 8 - 9 4149 147
XM_017002326.1 XP_016857815.1 3 - 4 3529 147

View Full Product Details