The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015904.3 | NP_056988.3 | 15 - 16 | 2583 | 109 |
GenBank mRNA | AB018284.1 | - | 15 - 16 | 2549 | 109 |
AF078035.1 | - | 15 - 16 | 2516 | 109 | |
AJ006776.1 | - | 15 - 16 | 2539 | 109 | |
AK001386.1 | - | 6 - 7 | 821 | 109 | |
AK308176.1 | - | 15 - 16 | 2565 | 109 | |
AL133563.1 | - | 12 - 13 | 2135 | 109 | |
BC032639.1 | - | 15 - 16 | 2505 | 109 |