Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF5B
Entrez Gene ID
9669
Gene Name
eukaryotic translation initiation factor 5B
Gene Aliases
IF2
Chromosome Location
Chr.2: 99337371 - 99400265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015904.3 NP_056988.3 3 - 4 429 105
GenBank mRNA AB018284.1 - 3 - 4 395 105
AF078035.1 - 3 - 4 362 105
AJ006776.1 - 3 - 4 385 105
AK308175.1 - 3 - 4 411 105
AK308176.1 - 3 - 4 411 105
BC006970.1 - 3 - 4 279 105
BC022260.1 - 3 - 4 307 105
BC032639.1 - 3 - 4 351 105
BX485839.1 - 3 - 4 429 105

View Full Product Details