The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005813.4 | NP_005804.1 | 2 - 3 | 984 | 66 |
XM_005264237.3 | XP_005264294.1 | 3 - 4 | 1257 | 66 | |
XM_005264238.3 | XP_005264295.1 | 2 - 3 | 233 | 66 | |
XM_017003743.1 | XP_016859232.1 | 1 - 2 | 141 | 66 | |
GenBank mRNA | AB015982.2 | - | 2 - 3 | 984 | 66 |
BC012820.2 | - | 2 - 3 | 943 | 66 | |
BC030706.2 | - | 2 - 3 | 929 | 66 |