Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5C1B
Entrez Gene ID
93034
Gene Name
5'-nucleotidase, cytosolic IB
Gene Aliases
AIRP, CN-IB, CN1B
Chromosome Location
Chr.2: 18562871 - 18589580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002006.2 NP_001002006.1 7 - 8 1312 152
NM_001199086.1 NP_001186015.1 7 - 8 1261 152
NM_001199087.1 NP_001186016.1 7 - 8 1363 152
NM_001199088.1 NP_001186017.1 7 - 8 1318 152
NM_033253.3 NP_150278.2 6 - 7 1132 152
GenBank mRNA AF352326.1 - 2 - 3 270 152
AF356185.1 - 6 - 7 1066 152
AF417165.1 - 7 - 8 1278 152
AJ295254.1 - 2 - 3 228 152
AK093234.1 - 3 - 4 415 152
AK093464.1 - 6 - 7 1057 152
AK097422.1 - 6 - 7 1121 152
AK302200.1 - 7 - 8 1207 152
AK302795.1 - 7 - 8 1347 152
BC136387.1 - 6 - 7 1085 152
BC171761.1 - 6 - 7 1085 152

Gene Symbol
NT5C1B-RDH14
Entrez Gene ID
100526794
Gene Name
NT5C1B-RDH14 readthrough
Gene Aliases
-
Chromosome Location
Chr.2: 18554723 - 18589580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199103.1 NP_001186032.1 6 - 7 1138 152
NM_001199104.1 NP_001186033.1 7 - 8 1312 152
GenBank mRNA AK057444.1 - 7 - 8 1294 152
AK307653.1 - 6 - 7 1122 152

View Full Product Details