The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270660.1 | NP_001257589.1 | 4 - 5 | 527 | 68 |
NM_001270661.1 | NP_001257590.1 | 2 - 3 | 356 | 68 | |
NM_005885.3 | NP_005876.2 | 5 - 6 | 671 | 68 | |
XM_011513932.2 | XP_011512234.1 | 5 - 6 | 429 | 68 | |
XM_011513934.1 | XP_011512236.1 | 5 - 6 | 632 | 68 | |
XM_011513936.2 | XP_011512238.1 | 5 - 6 | 632 | 68 | |
XM_017008944.1 | XP_016864433.1 | 5 - 6 | 632 | 68 | |
GenBank mRNA | AB011169.1 | - | 5 - 6 | 594 | 68 |
AK000970.1 | - | 5 - 6 | 586 | 68 | |
AK296585.1 | - | 2 - 3 | 279 | 68 | |
AK300034.1 | - | 4 - 5 | 463 | 68 | |
BC136461.1 | - | 5 - 6 | 543 | 68 | |
BC136462.1 | - | 5 - 6 | 543 | 68 | |
BC142679.1 | - | 5 - 6 | 522 | 68 | |
BC142694.1 | - | 5 - 6 | 522 | 68 |