The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012310.4 | NP_036442.3 | 9 - 10 | 1216 | 68 |
GenBank mRNA | AB208797.1 | - | 9 - 10 | 1116 | 68 |
AF071592.2 | - | 9 - 10 | 1133 | 68 | |
AF179308.1 | - | 8 - 9 | 1073 | 68 | |
AF277375.1 | - | 6 - 7 | 692 | 68 | |
AJ271784.1 | - | 8 - 9 | 1079 | 68 | |
AK022592.1 | - | 6 - 7 | 676 | 68 | |
AK313133.1 | - | 8 - 9 | 1449 | 68 | |
BC003664.2 | - | 9 - 10 | 1154 | 68 | |
BC038459.1 | - | 9 - 10 | 1153 | 68 | |
BC049218.1 | - | 9 - 10 | 1122 | 68 | |
BC050548.1 | - | 9 - 10 | 1115 | 68 |