Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
Chromosome Location
Chr.2: 84459588 - 84819589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370.1 NP_001361.1 56 - 57 9516 108
XM_006711956.2 XP_006712019.1 56 - 57 9621 108
XM_011532649.2 XP_011530951.1 56 - 57 9470 108
XM_011532650.2 XP_011530952.1 56 - 57 9471 108
XM_011532652.1 XP_011530954.1 55 - 56 9390 108
XM_011532653.1 XP_011530955.1 55 - 56 9369 108
XM_011532654.1 XP_011530956.1 55 - 56 9249 108
XM_011532655.2 XP_011530957.1 52 - 53 8538 108
XM_011532657.1 XP_011530959.1 50 - 51 8277 108
XM_017003521.1 XP_016859010.1 54 - 55 9243 108
XM_017003522.1 XP_016859011.1 51 - 52 9342 108
XM_017003523.1 XP_016859012.1 51 - 52 8509 108
GenBank mRNA AB051484.2 - 20 - 21 3451 108
AB290165.1 - 22 - 23 3655 108
BC117259.1 - 9 - 10 1648 108

View Full Product Details