Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
Chromosome Location
Chr.2: 84459588 - 84819589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370.1 NP_001361.1 25 - 26 4090 100
XM_006711956.2 XP_006712019.1 25 - 26 4195 100
XM_011532649.2 XP_011530951.1 25 - 26 4044 100
XM_011532650.2 XP_011530952.1 25 - 26 4045 100
XM_011532652.1 XP_011530954.1 25 - 26 4090 100
XM_011532653.1 XP_011530955.1 25 - 26 4090 100
XM_011532654.1 XP_011530956.1 24 - 25 3823 100
XM_011532655.2 XP_011530957.1 21 - 22 3112 100
XM_011532657.1 XP_011530959.1 19 - 20 2851 100
XM_011532660.1 XP_011530962.1 25 - 26 4090 100
XM_017003521.1 XP_016859010.1 25 - 26 4090 100
XM_017003522.1 XP_016859011.1 20 - 21 3916 100
XM_017003523.1 XP_016859012.1 20 - 21 3083 100
XM_017003524.1 XP_016859013.1 25 - 26 4090 100

View Full Product Details