Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGAT1
Entrez Gene ID
8694
Gene Name
diacylglycerol O-acyltransferase 1
Gene Aliases
ARAT, ARGP1, DGAT, DIAR7
Chromosome Location
Chr.8: 144314583 - 144326919 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012079.5 NP_036211.2 12 - 13 1264 70
XM_011517356.2 XP_011515658.1 12 - 13 985 70
GenBank mRNA AB057815.1 - 12 - 13 1007 70
AF059202.1 - 12 - 13 1225 70
BC006263.2 - 11 - 12 717 70
BC015762.1 - 12 - 13 1229 70
BC023565.1 - 12 - 13 1234 70
BC150649.1 - 12 - 13 1210 70

View Full Product Details