The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006391.2 | NP_006382.1 | 14 - 15 | 1728 | 83 |
GenBank mRNA | AF098799.1 | - | 14 - 15 | 1693 | 83 |
AK074908.1 | - | 7 - 8 | 759 | 83 | |
AK312885.1 | - | 14 - 15 | 1699 | 83 | |
BC114929.1 | - | 14 - 15 | 1654 | 83 |